Learn More
Waters Corp Solvent Bottle Tray WAT271082
Supplier:  Waters Corp WAT271082
Encompass_Preferred
Replacement Solvent Bottle Tray. The Solvent Bottle Tray holds four 1-Liter bottles. The solvents should be placed above the gradient proportioning valve. This tray was used as the standard on Alliance Separation Modules for many years and is now an option on current Alliance Separation Modules.
Catalog No. 50-793-841