Learn More
MSC Rite-Kem Inc Organic Solvent Degreaser, Halogen-Free, 12 x 1 qt
Catalog No. 502838563
Encompass
This organic solvent degreaser is designed for light to medium duty cleaning of various metal surfaces.
  • Superior formulated organic solvent degreaser
  • Suitable for light to medium duty cleaning
  • Effective on various metal surfaces
  • Halogen-free
  • Packaged in 12 x 1 qt containers
Catalog No. 50-283-8563 Supplier MSC Supplier No. 75311274
This organic solvent degreaser is designed for light to medium duty cleaning of various metal surfaces.
  • Superior formulated organic solvent degreaser
  • Suitable for light to medium duty cleaning
  • Effective on various metal surfaces
  • Halogen-free
  • Packaged in 12 x 1 qt containers