Learn More
Mac-Mod SafetyCap II, 45 mm, 2x PFA-fitting 3.2 mm OD + air valve
SDP
Catalog No. 5000151604
Encompass
MAC-MOD provides perfect protection from hazardous vapours, reduction of expensive solvent losses, guarantee of pure eluents and easy handling of hazardous chemicals, with a full range of safety products that can be utilized to ensure your dangerous chemicals that you handle on a daily basis, stay enclosed where they belong.
Catalog No. 50-001-51604 Supplier Mac-Mod Supplier No. MMBS452C
Add to Cart
Add to Cart
MAC-MOD provides perfect protection from hazardous vapours, reduction of expensive solvent losses, guarantee of pure eluents and easy handling of hazardous chemicals, with a full range of safety products that can be utilized to ensure your dangerous chemicals that you handle on a daily basis, stay enclosed where they belong.