Learn More
Mac-Mod SafetyWasteCap for 45 mm cap with 4x 2,3/3,2 mm OD tubing connections, 1x 6,4 - 9 mm ID fitting + connector for exhaust filter
SDP
Catalog No. 5000150923
Encompass
MAC-MOD provides perfect protection from hazardous vapours, reduction of expensive solvent losses, guarantee of pure eluents and easy handling of hazardous chemicals, with a full range of safety products that can be utilized to ensure your dangerous chemicals that you handle on a daily basis, stay enclosed where they belong.
Catalog No. 50-001-50923 Supplier Mac-Mod Supplier No. MMBS45W4C1T
MAC-MOD provides perfect protection from hazardous vapours, reduction of expensive solvent losses, guarantee of pure eluents and easy handling of hazardous chemicals, with a full range of safety products that can be utilized to ensure your dangerous chemicals that you handle on a daily basis, stay enclosed where they belong.