Learn More
Ethos Biosciences Reagent Alcohol 100%, 1gal, Ethos
Supplier:  Ethos Biosciences 3335G
Encompass_Preferred
Reagent Alcohol 100% is a high-purity alcohol blend suitable for pathology, histology, cytology, and general laboratory applications. Commonly used as a dehydrating agent, fixative, and solvent, it is ideal for tissue processing, slide preparation, and staining protocols. This clear, colorless solution provides consistent performance for reliable specimen preparation and analytical results. It is also widely used for cleaning, reagent preparation, and general laboratory tasks. Supplied in chemically compatible containers, Reagent Alcohol 100% is appropriate for clinical, research, pharmaceutical, and industrial laboratories.
Catalog No. 50-005-74312