Learn More
Ethos Biosciences Ethanol 70%, Denatured, 1gal, Ethos
Catalog No. 5000574341
Encompass_Preferred
Ethanol 70% Denatured is a solvent suitable for general laboratory, pathology, and research applications. Commonly used as a dehydrating agent in tissue processing, it is ideal for histology workflows, slide preparation, and reagent formulation. This clear, colorless alcohol provides consistent performance for reliable specimen handling and analytical results. It is also widely used for cleaning, solvent extraction, instrument maintenance, and other routine laboratory tasks. Supplied in chemically compatible containers, Ethanol 70% Denatured is appropriate for clinical, research, pharmaceutical, and industrial laboratory environments.
Catalog No. 50-005-74341 Supplier Ethos Biosciences Supplier No. 3337G
Ethanol 70% Denatured is a solvent suitable for general laboratory, pathology, and research applications. Commonly used as a dehydrating agent in tissue processing, it is ideal for histology workflows, slide preparation, and reagent formulation. This clear, colorless alcohol provides consistent performance for reliable specimen handling and analytical results. It is also widely used for cleaning, solvent extraction, instrument maintenance, and other routine laboratory tasks. Supplied in chemically compatible containers, Ethanol 70% Denatured is appropriate for clinical, research, pharmaceutical, and industrial laboratory environments.