Learn More
Perkin Elmer US LLC Solvent Flex Two-Stop Peristaltic Pump Tubing - 0.38 mm I.D., Orange/Green, Pkg. 12
Catalog No. 502112238
Encompass_Preferred
This Solvent Flex Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. 50-211-2238 Supplier Perkin Elmer US LLC Supplier No. N0777874
Add to Cart
Add to Cart
This Solvent Flex Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.