Learn More
Chemglass Life Sciences Solvent Guide (Gregar Extractor)
SDP
Catalog No. 50873065
Encompass_Preferred
Solvent guide for the Gregar Extractor (CG137501). During solid extractions the extractor operates upon the principle of continuous solvent displacement, permitting the sample to be immersed in solvent at all times, thereby increasing the efficiency of the solid-solvent interactions. The design eliminates packing and/or channeling of solid samples normally associated with common soxhlet extractors. For more information, please refer to item CG137501.
Catalog No. 50-873-065 Supplier Chemglass Life Sciences Supplier No. CG137505
Solvent guide for the Gregar Extractor (CG137501). During solid extractions the extractor operates upon the principle of continuous solvent displacement, permitting the sample to be immersed in solvent at all times, thereby increasing the efficiency of the solid-solvent interactions. The design eliminates packing and/or channeling of solid samples normally associated with common soxhlet extractors. For more information, please refer to item CG137501.