Learn More
Waters Corp Waters Solvent Inlet Filter, 10 μm, 1/8 in. OD
Supplier:  Waters Corp PSL613578
Encompass_Preferred
The Solvent Inlet Filter 10 μm 1/8 in. protects high-performance liquid chromatography (HPLC) systems from harmful particulate matter. It has a precise pore size of 10 μm to eliminate damaging particles and a 1/8 in. outer diameter for compatibility with various HPLC systems. This filter is applicable in chromatography, liquid handling, sample preparation, and general laboratory use to prevent interference with chromatography results and protect equipment from damage or contamination.
  • Pore size of 10 μm for effective particle removal.
  • Outer diameter of 1/8 in. ensures seamless integration with various HPLC systems.
  • Constructed from durable and corrosion-resistant stainless steel.
  • Precision-machined threads for a secure connection.
  • Effortless installation and user-friendly.
Catalog No. 50-784-248