Learn More
Waters Corp WATERS CORPORATION Solvent Inlet Filter, 10μm, 1/8in OD
Supplier:  Waters Corp PSL613609
Encompass_Preferred
This solvent inlet filter protects analytical instruments from particulate contamination. It is a chemically inert 10μm filter compatible with various solvents, extending instrument lifespan and improving accuracy and reproducibility of results.
  • Crafted from chemically inert materials like PTFE or stainless steel for compatibility with diverse solvents in LC, GC, sample preparation, storage, and transport applications.
  • Features a 10μm filter size, preventing particulate contamination from reaching lab equipment.
  • Safeguards analytical instruments, extending their lifespan and reducing maintenance needs.
  • Enhances accuracy and reproducibility of analytical results by preventing contamination issues.
  • Compatible with a range of liquid chromatography products.
  • Ensures solvents pass through seamlessly without compromising sample purity.
Catalog No. 50-784-254