Learn More
Waters Corp Solvent Inlet Manifold WAS207074
Catalog No. 50793470 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Solvent Inlet Manifold provides the connection for eluent inlet tubing and routes eluent to the inlet check valve on each pump head.
Catalog No. 50-793-470 Supplier Waters Corp Supplier No. WAS207074
The Solvent Inlet Manifold provides the connection for eluent inlet tubing and routes eluent to the inlet check valve on each pump head.