Learn More
Ethos Biosciences Acetone, 16oz, Ethos
Catalog No. 5000574229
Encompass_Preferred
Acetone is a high-purity solvent suitable for pathology, histology, cytology, and general laboratory applications. Commonly used as a tissue fixative, dehydrating agent, and slide preparation solvent, it is ideal for immunohistochemistry, frozen section procedures, and rapid fixation protocols. This clear, colorless, fast-evaporating solvent delivers consistent performance for reliable specimen processing and microscopic evaluation. It is also widely used for general laboratory tasks including cleaning, degreasing, solvent extraction, and reagent preparation. Supplied in chemically compatible containers, acetone is appropriate for clinical, research, and industrial laboratories requiring a versatile, high-quality solvent.
Catalog No. 50-005-74229 Supplier Ethos Biosciences Supplier No. 330516
Acetone is a high-purity solvent suitable for pathology, histology, cytology, and general laboratory applications. Commonly used as a tissue fixative, dehydrating agent, and slide preparation solvent, it is ideal for immunohistochemistry, frozen section procedures, and rapid fixation protocols. This clear, colorless, fast-evaporating solvent delivers consistent performance for reliable specimen processing and microscopic evaluation. It is also widely used for general laboratory tasks including cleaning, degreasing, solvent extraction, and reagent preparation. Supplied in chemically compatible containers, acetone is appropriate for clinical, research, and industrial laboratories requiring a versatile, high-quality solvent.