Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solvent Oils and Fats, AQUASTAR™, MilliporeSigma™

Catalog No. 50947570 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50947570 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947570 Supplier MilliporeSigma Supplier No. 1880161000/DEL

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Solvent Oils and Fats
Name Note AQUASTAR™, Solvent for volumetric Karl Fischer titration with two component reagents for oils and fats
Quantity 1 L
Synonym Solvent, Karl Fischer Volumetric Titration, Oils and Fats
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.