Learn More
Chemglass Life Sciences Solvent Pickup Adapter, GL45 to GPI 38 Thread
SDP
Catalog No. 501311011
Encompass_Preferred
Replacement solvent pickup adapter for the CG1167 series solvent reservoir.
Catalog No. 50-131-1011 Supplier Chemglass Life Sciences Supplier No. CG116740
Replacement solvent pickup adapter for the CG1167 series solvent reservoir.