Learn More
BURKLE INC Solvent pump hand operated, 23.62 in. (60 cm), G 2"
Catalog No. 5000026094
Encompass_Preferred
Hand-operated solvent pump for contaminationfree removal of organic solvents and flavouring agents. The hand pump works according to the overpressure principle. An excess pressure is built up in the tank with the pump ball, forcing the liquid out of the container.
Pouring flammable liquids
All medium-contacting parts are made of stainless steel AISI 304 (1.4301) and PTFE. The Bürkle anti-static set can be used to ground the pump for dispensing flammable liquids so that no electrostatic charge forms.
Gastight version
The pump can be completely closed off with the two ball valves so that no dangerous vapours can escape after dispensing.
Suitable for different containers
The hand pump is suitable for containers up to 60 litres.
Catalog No. 50-000-26094 Supplier BURKLE INC Supplier No. 56031000
Hand-operated solvent pump for contaminationfree removal of organic solvents and flavouring agents. The hand pump works according to the overpressure principle. An excess pressure is built up in the tank with the pump ball, forcing the liquid out of the container.
Pouring flammable liquids
All medium-contacting parts are made of stainless steel AISI 304 (1.4301) and PTFE. The Bürkle anti-static set can be used to ground the pump for dispensing flammable liquids so that no electrostatic charge forms.
Gastight version
The pump can be completely closed off with the two ball valves so that no dangerous vapours can escape after dispensing.
Suitable for different containers
The hand pump is suitable for containers up to 60 litres.