Learn More
SPECTRUM QUALITY STANDARDS Solvent Red 164 Dye in Kerosene, Custom Daily Check Standard, 100 mL
Catalog No. NC1031633
Encompass_Preferred
This standard is used for the determination of dye Solvent Red 164 in commercially available Diesel Fuels using Visible Spectroscopy. The Standards are made with Solvent Red 26, which is available in a Certified Pure form; against which, the Solvent Red 164 is measured.
Catalog No. NC1031633 Supplier SPECTRUM QUALITY STANDARDS Supplier No. 6258DCK
This standard is used for the determination of dye Solvent Red 164 in commercially available Diesel Fuels using Visible Spectroscopy. The Standards are made with Solvent Red 26, which is available in a Certified Pure form; against which, the Solvent Red 164 is measured.