Learn More
Chemglass Life Sciences Solvent Reservoir System, 10L, Complete
SDP
Catalog No. 501311004
Encompass_Preferred
Complete 10L Solvent Reservoir Consists of:1 Plastic Coated Glass Reservoir1 3-Hole Cap Assembly1 Combination Inlet/Sparger Filter, 2um2 CG1164N02 Flangeless Nuts for 1/8” Tubing2 CG1164F02 Flangeless Ferrule for 1/8” Tubing1 CG116610 Tefzel Plug1 CG116402 1/8” PTFE Tubing x 10’ long The bottle is blue tinted plastic coated flat bottom, which provides additional safety by containing the glass fragments from accidents.A CG116745 cap assembly, manufactured of Polybutyleneteraphthalate (PBT) & PTFE will allow the cap to withstand higher temperatures 200C° & is resistant to alcohols, ethers, hydrocarbons, & strong acids. The CG116608 cap assembly fits GL45 threaded bottles & has a PTFE bulkhead w/ three 1/4"- 28 threaded ports to accommodate fittings for PTFE 1/8" O.D. tubing.The bottom of the bottle inlet filter has a 2um filter which provides sparging capabilities preventing bubbles from entering the system.NOTE: The 2µm filter has a maximum flow rate of 30mL per minute.
Catalog No. 50-131-1004 Supplier Chemglass Life Sciences Supplier No. CG116714
Complete 10L Solvent Reservoir Consists of:1 Plastic Coated Glass Reservoir1 3-Hole Cap Assembly1 Combination Inlet/Sparger Filter, 2um2 CG1164N02 Flangeless Nuts for 1/8” Tubing2 CG1164F02 Flangeless Ferrule for 1/8” Tubing1 CG116610 Tefzel Plug1 CG116402 1/8” PTFE Tubing x 10’ long The bottle is blue tinted plastic coated flat bottom, which provides additional safety by containing the glass fragments from accidents.A CG116745 cap assembly, manufactured of Polybutyleneteraphthalate (PBT) & PTFE will allow the cap to withstand higher temperatures 200C° & is resistant to alcohols, ethers, hydrocarbons, & strong acids. The CG116608 cap assembly fits GL45 threaded bottles & has a PTFE bulkhead w/ three 1/4"- 28 threaded ports to accommodate fittings for PTFE 1/8" O.D. tubing.The bottom of the bottle inlet filter has a 2um filter which provides sparging capabilities preventing bubbles from entering the system.NOTE: The 2µm filter has a maximum flow rate of 30mL per minute.