Learn More
Description
- Ideal for lab applications where durability to environmental and chemical conditions is key
- Designed to withstand extreme temperatures and short-term exposure to chemicals such as xylene, DMSO and ethanol
- Made with a permanent acrylic adhesive for secure bonding to flat, smooth surfaces such as stainless steel and polypropylene
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | UL Recognized to UL969, CSA Accepted |
| Adhesive Type | Permanent Acrylic |
| Coating | None |
| Color | White |
| Finish | Matte |
| Format | Die-cut |
| Material | Polyester |
| Printable Sides | Top |
| Print Technology | Thermal Transfer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.