Learn More
Uline SOLVENT RESISTANT SPRAY NOZZLE
Catalog No. NC2379426
Encompass
Solvent Resistant Spray Nozzle 3/packSuper-charged nozzle sprays three times more than standard nozzles.For use with acetone and solvent-based chemicals.Leakproof, solvent resistant brass nozzle.
Catalog No. NC2379426 Supplier Uline Supplier No. S23427PK
Solvent Resistant Spray Nozzle 3/packSuper-charged nozzle sprays three times more than standard nozzles.For use with acetone and solvent-based chemicals.Leakproof, solvent resistant brass nozzle.