Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Solvent-Resistant White Biopsy Bags

Catalog No. 6774024 Shop All Epredia Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
67-740-24 Large
67-740-22 Small
67-740-19 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 67-740-24 Supplier Epredia™ Supplier No. 6774024
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solvent-resistant nylon and thin mesh provide, and allow for optimal fluid exchange around the tissue

  • Minimize the risk of small specimen loss
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Rapidly remove specimens with convenient peel-open bag

Specifications

Product Type Biopsy Bag
Material Nylon, Solvent-resistant
Color White
Length (Metric) 75 mm
Width (Metric) 85 mm
Length (English) 3 in.
Width (English) 3.5 in.
Dimensions (L x W) 3.0 x 3.5 in.
Size Large
Quantity 500/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.