Learn More
Sigma Aldrich Fine Chemicals Biosciences Solvent SafeTM pipette tip
Catalog No. 502900450
Encompass
Solvent Safe carbon filter pipette tipsfilters block corrosive vapors created when pipetting volatile organic solvents such as TfASafely and accurately pipette phenol/chloroform DNA extractionsPerform peptide synthesis and HPLC analysisExcellent alternative to glass pipettes
Catalog No. 50-290-0450 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S80641PAK
Solvent Safe carbon filter pipette tipsfilters block corrosive vapors created when pipetting volatile organic solvents such as TfASafely and accurately pipette phenol/chloroform DNA extractionsPerform peptide synthesis and HPLC analysisExcellent alternative to glass pipettes