Learn More
Synthware™ Solvent Still with Built in Condenser
GREENER_CHOICESDP
Catalog No. 31501856 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
125 mL
Catalog No. Capacity (Metric)
31-501-856 125 mL
1 options
Catalog No. 31-501-856 Supplier Synthware™ Supplier No. S144125
Add to Cart
Add to Cart
Grease Free Repurification Solvent Still Head
Specifications
Includes PTFE Stopper and Nylon Compression Cap
For Use With (Application) Organic Synthesis
Type Grease-free
Material Borosilicate Glass
Capacity (Metric) 125 mL
Quantity 1 Ea.
Size 24/40