Learn More
Synthware™ Solvent Still Head with Drip Tip
GREENER_CHOICESDP
Catalog No. 31501851 Shop All Kemtech Products
Change view
Click to view available options
Size:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-501-851 500 mL
31-501-850 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-501-851 Supplier Synthware™ Supplier No. S234004
Add to Cart
Add to Cart
Grease free, easy syringe access, and easy drainage
Grease free solvent still head with a drip tip on the delivery port. Also features 4 mm stopcocks on the upper portion for easy syringe needle access.
Specifications
Includes PTFE Plug and T-Bore Stopcock
For Use With (Application) Organic Synthesis
Type Solvent Still Head
Material Borosilicate Glass
Quantity 1 Ea.
Joint Type Ground Glass, 24/20
Size 500 mL