Learn More
Synthware™ Solvent Transfer Manifold
GREENER_CHOICESDP
Catalog No. 31501807 Shop All Kemtech Products
Change view
Click to view available options
Material:
Borosilicate Glass
Catalog No. Material
31-501-807 Borosilicate Glass
1 options
Catalog No. 31-501-807 Supplier Synthware™ Supplier No. M164380
Add to Cart
Add to Cart
For purifying and transferring solvents under inert conditions
Specifications
Type Solvent Transfer Manifold
For Use With (Application) Organic Synthesis
Includes High Vacuum Valves
Material Borosilicate Glass
Width (Metric) 380 mm
Height (Metric) 140 mm