Learn More
Chemglass Life Sciences Solvent Transfer Manifold, 24/40, Airfree, Schlenk
SDP
Catalog No. AF055801
Encompass_Preferred
Solvent Transfer Manifold, 24/40, Airfree, Schlenk. Used to purify and transfer solvents under inert conditions. Permits transfer of solvents directly into an evacuated storage vessel with 24/40 standard taper outer joints. A 0-4mm Chem-Vac™ Chem-Cap valve is used for vacuum control.
Catalog No. AF055801 Supplier Chemglass Life Sciences Supplier No. AF055801
Solvent Transfer Manifold, 24/40, Airfree, Schlenk. Used to purify and transfer solvents under inert conditions. Permits transfer of solvents directly into an evacuated storage vessel with 24/40 standard taper outer joints. A 0-4mm Chem-Vac™ Chem-Cap valve is used for vacuum control.