Learn More
Medchemexpress LLC Solvent violet 9 | 467-63-0 | MFCD00071920 | 389.53 g/mol | C25H31N3O | 10G
SDP
Catalog No. 5000239087
Encompass_Preferred
Solvent violet 9 is a triarylmethane dye used as a DNA stain in biochemical and molecular biology applications. It appears as a light blue to blue solid and is employed for nucleic acid visualization in research and diagnostic workflows. Chemical formula C25H31N3O, molecular weight 389.53 g/mol.
Catalog No. 50-002-39087 Supplier Medchemexpress LLC Supplier No. HY12578610G
Solvent violet 9 is a triarylmethane dye used as a DNA stain in biochemical and molecular biology applications. It appears as a light blue to blue solid and is employed for nucleic acid visualization in research and diagnostic workflows. Chemical formula C25H31N3O, molecular weight 389.53 g/mol.