Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Solvent Yellow 93 | 4702-90-3 | 99.1% | 25 MG
SDP

Catalog No. 5000413591
Encompass_Preferred

Solvent Yellow 93 (C.I. 48160) is an azomethine dye used as a colorant for toner and for coloring polyester fiber pulp, also used to make polyester masterbatch. It is for research use only.

  • Molecular weight: 358.39
  • Formula: C21H18N4O2
  • Appearance: Solid
  • Color: Yellow to orange
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Catalog No. 50-004-13591 Supplier Medchemexpress LLC Supplier No. 25MGHYD0376
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solvent Yellow 93 (C.I. 48160) is an azomethine dye used as a colorant for toner and for coloring polyester fiber pulp, also used to make polyester masterbatch. It is for research use only.

  • Molecular weight: 358.39
  • Formula: C21H18N4O2
  • Appearance: Solid
  • Color: Yellow to orange
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.