Learn More
Medchemexpress LLC Solvent Yellow 93 (C.I. 48160) | 4702-90-3 | 99.1% | 100 MG
SDP
Catalog No. 5000410246
Encompass_Preferred
Solvent Yellow 93 (C.I. 48160) is an azomethine dye characterized by its yellow to orange appearance and high purity. It is primarily used as a versatile colorant in various industrial applications.
  • Used as a colorant for toners
  • Suitable for coloring polyester fiber pulp
  • Applied in the production of polyester masterbatch
Catalog No. 50-004-10246 Supplier Medchemexpress LLC Supplier No. HYD0376100MG
Solvent Yellow 93 (C.I. 48160) is an azomethine dye characterized by its yellow to orange appearance and high purity. It is primarily used as a versatile colorant in various industrial applications.
  • Used as a colorant for toners
  • Suitable for coloring polyester fiber pulp
  • Applied in the production of polyester masterbatch