Learn More
Bellco Glass, Inc. Somatic Cell Slide,25x75mm
SDP
Catalog No. 501945824
Encompass_Preferred
The somatic Cell Count Slide is useful for direct microscopic counts of somatic cells in cows milk. Spread 0.01mL sample onto each circle than dry and stain. Count cells under oil immersion microscopy. Measurement: 25x75cm. MADE IN THE USA
Catalog No. 50-194-5824 Supplier Bellco Glass, Inc. Supplier No. 563801930
The somatic Cell Count Slide is useful for direct microscopic counts of somatic cells in cows milk. Spread 0.01mL sample onto each circle than dry and stain. Count cells under oil immersion microscopy. Measurement: 25x75cm. MADE IN THE USA