Learn More
American Radiolabeled Chemicals Inc SOMATOSTATIN 125I 50UC
SDP
Catalog No. 50901086
Encompass
Somatostatin-125I 50uCi in buffered solution (pH 7.2), 0.1 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1778
Catalog No. 50-901-086 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 017950UC
Add to Cart
Add to Cart
Request a Quote
Somatostatin-125I 50uCi in buffered solution (pH 7.2), 0.1 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1778