Learn More
Thermo Scientific Chemicals Somatostatin 14
Description
Somatostatin is used as a growth hormone-release inhibiting factor, treatment of severe, acute hemorrhage of gastroduodenal ulcers, experimental antidiabetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Purification Method | HPLC |
| Common Name | Somatostatin |
| Content And Storage | -30°C to -10°C |
| Format | Powder |
| Molecular Weight (g/mol) | 1637.89 |
| Product Type | Somatostatin 14 |
| Quantity | 5 mg |
| Regulatory Status | RUO |
| Sequence | Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys, (Disulfide bridge: Cys3-Cys14) |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.