Learn More
Eurofins Discoverx Somatostatin-28, 0.5 mg. (92-1068)
Supplier:  Eurofins Discoverx 921068
Encompass
Somatostatin 28 is an active fragment of somatostatin, an inhibitory peptide hormone produced in the hypothalamus that interacts with G protein-coupled somatostatin receptors to regulate the endocrine system, neurotransmission, and cell proliferation. Acts as an agonist on SSTR1-5 to inhibit release of insulin, glucagon, and numerous secondary hormones as well as regulate voltage-dependent calcium channels. Somatostatin 28 may also inhibit secretion of peptide YY. Provided as 0.5 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9608