Learn More
Bachem DOTA-(Tyr3)-Octreotide Acetate
SDP
Supplier:  Bachem 4043535.0001
Encompass_Preferred
DOTA-(Tyr3)-Octreotide Acetate, 4,7,10-Tricarboxymethyl-1,4,7,10-tetraaza-cyclododecan-1-yl-acetyl-D-Phe-Cys-Tyr-D-Trp-Lys-Thr-Cys-L-threoninol, 204318-14-9, C65H92N14O18S2, Mr 1421.66, 1mg. DOTATOC, Edotreotide. Somatostatin analog for tumor scintigraphy and radionuclide therapy.
Catalog No. 50-194-7500