Learn More
Santa Cruz Biotechnology Anti-Somatostatin Antibody (G-10), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC55565
Encompass
Somatostatin Antibody (G-10) is recommended for detection of Somatostatin of human origin by WB, IP, IF, IHC(P) and ELISA.
Catalog No. NC1144838