Learn More
Bioss Somatostatin Receptor 1 Recomb
SDP
Catalog No. 5000065691
Encompass_Preferred
Somatostatin Receptor 1 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IF(ICC) RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-65691 Supplier Bioss Supplier No. BSM62545R
Somatostatin Receptor 1 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IF(ICC) RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol