Learn More
ABclonal Technology Somatostatin Receptor 2 SSTR2
SDP
Catalog No. 5000006546
Encompass_Preferred
Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins The biologic effects of somatostatin are probably mediated by a family of G protein-coupled receptors that are expressed in a tissue-specific manner SSTR2 is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in cerebrum and kidney
Catalog No. 50-000-06546 Supplier ABclonal Technology Supplier No. A2197020UL
Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins The biologic effects of somatostatin are probably mediated by a family of G protein-coupled receptors that are expressed in a tissue-specific manner SSTR2 is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in cerebrum and kidney