Learn More
ABclonal Technology Somatostatin Receptor 2 SSTR2
Supplier: ABclonal Technology A21970100UL
Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins The biologic effects of somatostatin are probably mediated by a family of G protein-coupled receptors that are expressed in a tissue-specific manner SSTR2 is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in cerebrum and kidney