Learn More
Cayman Chemical Somatotropin Long Isoform hu
SDP
Catalog No. 502949293
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged somatotropin expressed in E. coli • Amino acids: 27-217 • MW: 24.7 kDa
Catalog No. 50-294-9293 Supplier Cayman Chemical Supplier No. 3209050 UG
Source: Recombinant human N-terminal His-tagged somatotropin expressed in E. coli • Amino acids: 27-217 • MW: 24.7 kDa