Learn More
Cayman Chemical Isomeranzin, Purity: ≥98%, CAS: 1088-17-1, 10 mg
SDP
Catalog No. 502910579
Encompass_Preferred
A coumarin with antioxidant and anti-inflammatory activities; inhibits superoxide anion generation in, and elastase release from, cytochalasin B- and fMLP-stimulated isolated human neutrophils (IC50s = 3.89 and 4.33 µM, respectively); inhibits LPS-induced NO release in RAW 264.7 macrophages at 30 µM; improves survival in a mouse model of LPS-induced septic shock and decreases disease severity in a mouse model of DSS-induced colitis at 30 mg/kg
Catalog No. 50-291-0579 Supplier Cayman Chemical Supplier No. 3582710 MG
A coumarin with antioxidant and anti-inflammatory activities; inhibits superoxide anion generation in, and elastase release from, cytochalasin B- and fMLP-stimulated isolated human neutrophils (IC50s = 3.89 and 4.33 µM, respectively); inhibits LPS-induced NO release in RAW 264.7 macrophages at 30 µM; improves survival in a mouse model of LPS-induced septic shock and decreases disease severity in a mouse model of DSS-induced colitis at 30 mg/kg