Learn More
SOMNI Scientific™ Mobile Research Workstation
SDP
Catalog No. 01001104 Shop All Somni Scientific Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
01-001-104 Large
1 options
Catalog No. 01-001-104 Supplier SOMNI Scientific™ Supplier No. MW17015
Add to Cart
Add to Cart
Mobile Research Workstation that is a fully independent mobile anesthesia system. Includes AMD-3+ Multi-delivery system, EPS-3 Exposure Prevention, Uniflow nosecone, Large induction chamber and Oxygen concentrator. Large work space and room to add organizational accessories.
  • Oxygen concentrator and/or e-cylinder mounting options
  • Customizable to add organizational accessories
  • Top surface is the perfect medium for SOMNI's magnetic accessories
  • Space for procedures
  • Stable, 3 in. easy roll wheels
Specifications
For Use With (Application) Animal Inhalant Anesthesia
Size Large
Animal Mouse and Rat, Paw
Type Anesthesia Machine