Learn More
SOMNI Scientific™ Mobile Work Station with Magnetic Work Area
SDP
Catalog No. 01001107 Shop All Somni Scientific Products
Change view
Click to view available options
Size:
Medium
Catalog No. Size
01-001-107 Medium
1 options
Catalog No. 01-001-107 Supplier SOMNI Scientific™ Supplier No. MW17020
Add to Cart
Add to Cart
Request a Quote
Customizable, mobile workspace with 'H' style frame that is stable and fits into tight spaces. The top work surface is the ideal medium for magnetic accessories.
  • H style frame
  • Accommodates an O2 Concentrator or e-cylinder
Specifications
For Use With (Application) Animal Inhalant Anesthesia
Size Medium
Animal Mouse and Rat, Paw
Type Anesthesia Machine