Learn More
Med Vet International Somni Oxygen Concentrators and Accessories - 5L Concentrator, J0744SN, Each
Catalog No. 5000583958
Encompass_Preferred
OT-Elite 5 Oxygen Concentrator Features: • 0-5 lpm • 5-11 PSI Outlet Pressure • Standard DISS fitting • Floating Ball Style Flowmeter • Sound level ?43dBa • 25"W x 15"W x 12.5"D • 50 lbs • 1-year warranty • 110V AC
Catalog No. 50-005-83958 Supplier Med Vet International Supplier No. JOR744SN
OT-Elite 5 Oxygen Concentrator Features: • 0-5 lpm • 5-11 PSI Outlet Pressure • Standard DISS fitting • Floating Ball Style Flowmeter • Sound level ?43dBa • 25"W x 15"W x 12.5"D • 50 lbs • 1-year warranty • 110V AC