Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ SOMNI Scientific™ Chemtron Female Quick Disconct
SDP

Catalog No. 04777226 Shop All Somni Scientific Products
Change view
Click to view available options
Color:
Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
04-777-226 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-777-226 Supplier SOMNI Scientific™ Supplier No. QD7004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A popular option with a simple button press for quickly disconnecting oxygen supply hose from the corresponding male quick connect

  • Quickly connects and disconnects from the corresponding male quick connect.
  • Supplied with a 0.25 inch hose barb
  • Color coded green for oxygen
  • Order with a custom hose assembly

Specifications

For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) For Oxygen is Supplied with a 0.25 in. hose barb
Color Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.