Learn More
SOMNI Scientific™ SOMNI Scientific™ CO2 E-Tank Reg
SDP
Catalog No. 04777229 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-229 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-229 Supplier SOMNI Scientific™ Supplier No. RF20016
Add to Cart
Add to Cart
Attaches to an e in. size tank and supplies a constant flow of carbon dioxide up to 12 LPM
  • Attaches to a small e size tank and supplies a constant flow of carbon dioxide up to 12 LPM
  • Gas specific regulator for CO2
  • E-tank
  • Flows from 0-12 LPM
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Includes Flowmeter