Learn More
SOMNI Scientific™ SOMNI Scientific™ E-Tank Washer Brass and Viton
SDP
Catalog No. 04777227 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-227 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-227 Supplier SOMNI Scientific™ Supplier No. RW20015
Add to Cart
Add to Cart
Made of durable viton and brass to provide a lasting seal between the e-tank regulator yoke and the e-tank. A better seal equals less oxygen leaks
  • Provides a lasting seal between the e-tank regulator yoke and the e-tank
  • A better seal equals less oxygen leaks
  • Made of durable viton and brass
  • Sold individually
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Material Viton and Brass