Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ SOMNI Scientific™ Endo Adapter
SDP

Catalog No. 04777195 Shop All Somni Scientific Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
04-777-195 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-777-195 Supplier SOMNI Scientific™ Supplier No. AE8007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Common adapter used to connect anesthesia and waste gas systems

  • The 15 mm endo adapter is a clear, common adapter used to connect anesthesia and waste gas systems together
  • 6 mm end fits securely in 0.25 in. ID tubing
  • Applications include: Nosecone, induction chamber, WAG filter

Specifications

Diameter (English) 0.25 in.
For Use With (Application) Research Animal Inhalant Anesthesia
Dimensions (L x W) 6 x 15 mm
Color Clear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.