Learn More
SOMNI Scientific™ SOMNI Scientific™ Med Air Hose Assembly
SDP
Catalog No. 04777222 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-222 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-222 Supplier SOMNI Scientific™ Supplier No. HF10014
Add to Cart
Add to Cart
Provided with standard medical DISS Female threaded fittings on both ends The DISS fitting mates with the DISS Male on the anesthesia system
  • The yellow medical air hose assembly is provided with standard medical DISS Female threaded fittings on both ends
  • The DISS fitting mates with the DISS Male on the anesthesia system
  • Medical grade DISS Medical Air fittings
  • 6 feet long
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Length (English) 6 ft.