Learn More
SOMNI Scientific™ SOMNI Scientific™ Nosecone Diaphragms
SDP
Catalog No. 04777198 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-198 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-198 Supplier SOMNI Scientific™ Supplier No. NC19901
Add to Cart
Add to Cart
Custom cut latex diaphragms allow the user to make an appropriately sized opening for the animal to breathe
  • The Latex Nosecone Diaphragms are custom cut to perfectly fit the SOMNI Scientific NRB nosecone. 
  • Sized to fit the NRB
  • User determines appropriately sized opening for the animal
  • Sold as a pack of 50
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Material Latex
Quantity 50/Pk.