Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ SOMNI Scientific™ O2 Chemtron Male Quick Disconnct
SDP

Catalog No. 04777225 Shop All Somni Scientific Products
Change view
Click to view available options
Color:
Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
04-777-225 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-777-225 Supplier SOMNI Scientific™ Supplier No. QD7003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A popular option for quickly connecting oxygen supply hose to the corresponding female outlet or quick connect

  • Option for quickly connecting or disconnecting oxygen supply hose to the corresponding female outlet or quick connect.
  • Supplied with a 0.25 inch hose barb
  • Simple button press for removal
  • Color coded green for oxygen (US)
  • Order with a custom hose assembly

Specifications

For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) For Oxygen is Supplied with a 0.25 in. hose barb
Color Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.