Learn More
SOMNI Scientific™ SOMNI Scientific™ SOMNI Bain Circuit
SDP
Catalog No. 04777211 Shop All Somni Scientific Products
Change view
Click to view available options
Length (English):
60 in.
Catalog No. Length (English)
04-777-211 60 in.
1 options
Catalog No. 04-777-211 Supplier SOMNI Scientific™ Supplier No. BC13011
The tube within a tube design provides heat and moisture conservation with fresh gas entering the circuit at the patient end. GS elbow included. PVC, Phthalate and Latex Free
  • A tube within a tube design that provides heat and moisture conservation
  • The fresh gas enters the circuit at the patient end
  • PVC, Phthalate and Latex Free
  • GS elbow included
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Length (English) 60 in.